Gene Transcripts

Gene Symbol
Avpi1
Entrez Gene ID
69534
Gene Name
arginine vasopressin-induced 1
Gene Aliases
2310008N12Rik, AA410041, VIT32, esu, mVIT32
Chromosome Location
Chr. 19: 42123275 - 42128993 on Build GRCm38
UniGene ID
Mm.30060
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027106.4 NP_081382.1 2 371
GenBank mRNA AK009243.1 - 2 372
BC027646.1 - 2 405 1344413

View Full Product Details