Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175185.4 | NP_780394.1 | 4 | 991 | |
GenBank mRNA | AK088412.1 | - | 4 | 975 | |
AK150638.1 | - | 2 | 658 | ||
AK152392.1 | - | 2 | 658 | ||
AK162245.1 | - | 4 | 861 | ||
BC065074.1 | - | 4 | 907 | 6809862 |