Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029714.3 | NP_083990.2 | 3 | 396 | |
GenBank mRNA | AK006911.1 | - | 3 | 387 | |
AK077059.1 | - | 3 | 378 | ||
AK138347.1 | - | 3 | 396 | ||
BB614579.1 | - | 3 | 216 | ||
BC144969.1 | - | 3 | 375 | ||
BY724013.1 | - | 3 | 396 | ||
GQ225581.1 | - | 2 | 297 |