Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134091.2 | NP_598852.2 | 6 | 615 | |
GenBank mRNA | AK142314.1 | - | 6 | 584 | |
AK170806.1 | - | 6 | 582 | ||
AK185066.1 | - | 2 | 200 | ||
AK189808.1 | - | 2 | 200 | ||
AY382616.1 | - | 6 | 569 | ||
BC018197.1 | - | 6 | 575 | 4190407 | |
BC024122.1 | - | 6 | 575 | 5097970 | |
BC025561.1 | - | 6 | 553 | 5322980 |