Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139272.2 | NP_644678.2 | 9 | 983 | |
GenBank mRNA | AF348968.1 | - | 9 | 851 | |
AK046567.1 | - | 9 | 941 | ||
AK145122.1 | - | 9 | 861 | ||
AK149609.1 | - | 9 | 911 | ||
AK151536.1 | - | 9 | 774 | ||
BC007172.1 | - | 8 | 718 | ||
BC053063.1 | - | 8 | 826 | 5702572 | |
BC059818.1 | - | 9 | 928 | 5697789 |