Gene Transcripts

Gene Symbol
H2-M10.3
Entrez Gene ID
110696
Gene Name
histocompatibility 2, M region locus 10.3
Gene Aliases
5.3H
Chromosome Location
Chr. 17: 36365004 - 36368417 on Build GRCm38
UniGene ID
Mm.303091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201608.2 NP_963902.1 2 101
GenBank mRNA AF539599.1 - 2 95
AY211080.1 - 2 100

View Full Product Details