Gene Transcripts
- Gene Symbol
- JPX
- Entrez Gene ID
- 554203
- Gene Name
- JPX transcript, XIST activator (non-protein coding)
- Gene Aliases
- DCBALD06, ENOX, LINC00183, NCRNA00183
- Chromosome Location
- Chr. X: 73944324 - 74070384 on Build GRCh38
- UniGene ID
- Hs.648316
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056172.1 |
- |
5 |
1477 |
|
BC029480.1 |
- |
1 |
166 |
|
View Full Product Details