Gene Transcripts

Gene Symbol
JPX
Entrez Gene ID
554203
Gene Name
JPX transcript, XIST activator (non-protein coding)
Gene Aliases
DCBALD06, ENOX, LINC00183, NCRNA00183
Chromosome Location
Chr. X: 73944324 - 74070384 on Build GRCh38
UniGene ID
Hs.648316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK056172.1 - 5 1598
BC029480.1 - 1 287

View Full Product Details