Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027514.2 | NP_081790.1 | 2 | 398 | |
GenBank mRNA | AB195430.1 | - | 2 | 355 | |
AJ563420.1 | - | 2 | 286 | ||
AK014446.1 | - | 2 | 369 | ||
AK038747.1 | - | 2 | 369 | ||
AK040334.1 | - | 2 | 369 | ||
AK078344.1 | - | 2 | 365 | ||
AK080753.1 | - | 2 | 369 | ||
AK165873.1 | - | 2 | 388 | ||
BC013673.1 | - | 2 | 355 | 4188634 | |
BC032283.1 | - | 2 | 358 | 5363187 | |
BC138219.1 | - | 2 | 343 | 8861238 | |
BC138220.1 | - | 2 | 343 | 8861239 | |
BY276828.1 | - | 2 | 399 |