Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029017.2 | NP_083293.1 | 2 | 128 | |
GenBank mRNA | BC029173.1 | - | 108 | ||
BC038065.1 | - | 2 | 99 | ||
BC087953.1 | - | 2 | 122 | 30119240 | |
BY013327.1 | - | 2 | 128 | ||
CX122942.1 | - | 2 | 107 |