Gene Transcripts

Gene Symbol
Dcpp1
Entrez Gene ID
13184
Gene Name
demilune cell and parotid protein 1
Gene Aliases
BC005655, C86550, Dcpp, Dcpp-1, p20
Chromosome Location
Chr. 17: 23880876 - 23882853 on Build GRCm38
UniGene ID
Mm.371556
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019910.2 NP_064294.2 3 209
GenBank mRNA BC005655.1 - 206
BY112697.1 - 3 206
BY150555.1 - 3 209

View Full Product Details