Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297439.1 | NP_001284368.1 | 13 | 1692 | |
NM_001528.3 | NP_001519.1 | 12 | 1671 | ||
GenBank mRNA | AK314099.1 | - | 13 | 1650 | |
BC112190.1 | - | 12 | 1671 | 8327658 | |
BC112192.1 | - | 12 | 1683 | 8327660 | |
D14012.1 | - | 12 | 1605 |