Gene Transcripts

Gene Symbol
Mblac1
Entrez Gene ID
330216
Gene Name
metallo-beta-lactamase domain containing 1
Gene Aliases
Chromosome Location
Chr. 5: 138194314 - 138195621 on Build GRCm38
UniGene ID
Mm.19719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177878.3 NP_808546.1 1 760
GenBank mRNA AK049381.1 - 1 761
BC038925.1 - 1 741 5374241

View Full Product Details