Gene Transcripts

Gene Symbol
Ccdc79
Entrez Gene ID
320022
Gene Name
coiled-coil domain containing 79
Gene Aliases
4930532D21Rik, BB085179, Terb1
Chromosome Location
Chr. 8: 104446718 - 104509887 on Build GRCm38
UniGene ID
Mm.249775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_180958.3 NP_851289.2 4 329
GenBank mRNA AB775896.1 - 2 110
AK029754.1 - 4 330
BC113771.1 - 3 653
BC129814.1 - 3 654

View Full Product Details