Gene Transcripts

Gene Symbol
Prol1
Entrez Gene ID
17830
Gene Name
proline rich, lacrimal 1
Gene Aliases
Muc10
Chromosome Location
Chr. 5: 88317312 - 88328817 on Build GRCm38
UniGene ID
Mm.200411
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008644.2 NP_032670.2 3 210
GenBank mRNA AF247816.1 - 3 212
BC021158.1 - 3 214
BC021401.1 - 3 212 4166946
BC042902.1 - 3 208 4924369
U37531.1 - 3 210

View Full Product Details