Gene Transcripts

Gene Symbol
Coa3
Entrez Gene ID
52469
Gene Name
cytochrome C oxidase assembly factor 3
Gene Aliases
1810033A19Rik, Ccdc56, D11Ertd99e, HSPC009
Chromosome Location
Chr. 11: 101277970 - 101278948 on Build GRCm38
UniGene ID
Mm.52
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026618.2 NP_080894.1 1 164
GenBank mRNA AK003955.1 - 1 162
AK019025.1 - 1 166
BC026206.1 - 1 158 1265058

View Full Product Details