Gene Transcripts

Gene Symbol
Olfr9
Entrez Gene ID
18373
Gene Name
olfactory receptor 9
Gene Aliases
MOR269-3
Chromosome Location
Chr. 10: 128989914 - 128990852 on Build GRCm38
UniGene ID
Mm.377111
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146861.1 NP_667072.1 1 819
GenBank mRNA BC116994.1 - 1 884
BC119365.1 - 1 884 8734114

View Full Product Details