Gene Transcripts

Gene Symbol
ATP5D
Entrez Gene ID
513
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit
Gene Aliases
Chromosome Location
Chr. 19: 1241750 - 1244825 on Build GRCh38
UniGene ID
Hs.418668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001975.1 NP_001001975.1 4 502
NM_001687.4 NP_001678.1 4 502
GenBank mRNA BC002389.2 - 4 502
BC004426.1 - 4 502 2819749
BC018079.1 - 4 404 4798666
BC018817.2 - 4 502
CR542218.1 - 4 401
S87916.1 - 4 484
S87918.1 - 4 484
X63422.1 - 4 484
X63423.1 - 4 484

View Full Product Details