Gene Transcripts

Gene Symbol
Olfr15
Entrez Gene ID
18312
Gene Name
olfactory receptor 15
Gene Aliases
MOR256-17, OR3
Chromosome Location
Chr. 16: 3838975 - 3839913 on Build GRCm38
UniGene ID
Mm.377102
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008762.2 NP_032788.2 1 199
GenBank mRNA BC100679.1 - 1 197
BC100680.1 - 1 198
BC100685.1 - 1 198
BC115838.1 - 1 198

View Full Product Details