Gene Transcripts

Gene Symbol
Vmn1r83
Entrez Gene ID
171243
Gene Name
vomeronasal 1 receptor 83
Gene Aliases
V1rg8
Chromosome Location
Chr. 7: 12321208 - 12322128 on Build GRCm38
UniGene ID
Mm.377199
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134209.1 NP_598970.1 1 333
GenBank mRNA BC125369.1 - 1 380
BC138162.1 - 1 380 8861181

View Full Product Details