Gene Transcripts

Gene Symbol
STK17B
Entrez Gene ID
9262
Gene Name
serine/threonine kinase 17b
Gene Aliases
DRAK2
Chromosome Location
Chr. 2: 196133583 - 196176503 on Build GRCh38
UniGene ID
Hs.88297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004226.3 NP_004217.1 4 675
XM_011512168.2 XP_011510470.1 4 640
XM_011512169.1 XP_011510471.1 4 673
XM_011512170.1 XP_011510472.1 5 656
XM_011512171.2 XP_011510473.1 3 168
GenBank mRNA AB011421.1 - 4 612
AK312865.1 - 4 673
BC016040.1 - 4 614 4575012
CR542210.1 - 3 351
CR542225.1 - 3 351

View Full Product Details