Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000439.4 | NP_000430.3 | 6 | 887 | |
NM_001177875.1 | NP_001171346.1 | 6 | 545 | ||
GenBank mRNA | AB208874.1 | - | 6 | 563 | |
AK303888.1 | - | 6 | 545 | ||
BC130295.1 | - | 6 | 769 | 40146326 | |
BC136486.1 | - | 6 | 769 | 9020475 | |
M90753.1 | - | 6 | 857 | ||
X64810.1 | - | 6 | 837 |