Gene Transcripts

Gene Symbol
PRSS42
Entrez Gene ID
339906
Gene Name
protease, serine 42
Gene Aliases
TESSP2
Chromosome Location
Chr. 3: 46830404 - 46834095 on Build GRCh38
UniGene ID
Hs.585127
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182702.1 NP_874361.1 4 722
GenBank mRNA AJ544583.1 - 4 722

View Full Product Details