Gene Transcripts

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
Chromosome Location
Chr. 1: 64470793 - 64693058 on Build GRCh38
UniGene ID
Hs.443891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293274.1 NP_001280203.1 6 1196
NM_020925.2 NP_065976.2 7 917
XM_011541862.1 XP_011540164.1 6 1192
XM_011541863.1 XP_011540165.1 7 1341
GenBank mRNA AB046793.1 - 5 699
BC039301.1 - 7 903 4836806
BX537603.1 - 6 1195

View Full Product Details