Gene Transcripts

Gene Symbol
PKD1L3
Entrez Gene ID
342372
Gene Name
polycystin 1 like 3, transient receptor potential channel interacting
Gene Aliases
Chromosome Location
Chr. 16: 71929538 - 72000401 on Build GRCh38
UniGene ID
Hs.675286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181536.1 NP_853514.1 6, 7 974
XM_017023201.1 XP_016878690.1 6, 7 1397
XM_017023202.1 XP_016878691.1 6, 7 1397
XM_017023203.1 XP_016878692.1 6, 7 1397
XM_017023204.1 XP_016878693.1 6, 7 1397
GenBank mRNA AY164485.1 - 6, 7 974

View Full Product Details