Gene Transcripts
- Gene Symbol
- CD22
- Entrez Gene ID
- 933
- Gene Name
- CD22 molecule
- Gene Aliases
- SIGLEC-2, SIGLEC2
- Chromosome Location
- Chr. 19: 35329166 - 35347361 on Build GRCh38
- UniGene ID
- Hs.579691
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001185099.1 |
NP_001172028.1
|
10, 11 |
2026 |
|
NM_001185101.1 |
NP_001172030.1
|
9, 10 |
1759 |
|
NM_001278417.1 |
NP_001265346.1
|
10, 11 |
2118 |
|
NM_001771.3 |
NP_001762.2
|
11, 12 |
2290 |
|
GenBank mRNA |
AK225625.1 |
- |
10, 11 |
2008 |
|
AK225694.1 |
- |
11, 12 |
2272 |
|
AK301177.1 |
- |
11, 12 |
2153 |
|
BC109306.1 |
- |
10, 11 |
2072 |
40035425 |
X52785.1 |
- |
8, 9 |
1709 |
|
X59350.1 |
- |
11, 12 |
2262 |
|
View Full Product Details