Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003878.2 | NP_003869.1 | 2 | 449 | |
XM_011517623.2 | XP_011515925.1 | 2 | 200 | ||
GenBank mRNA | AK290450.1 | - | 2 | 193 | |
AK301094.1 | - | 2 | 199 | ||
BC025025.1 | - | 2 | 250 | 3924620 | |
U55206.1 | - | 2 | 225 |