Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004577.3 | NP_004568.2 | 6 | 1024 | |
XM_005271773.1 | XP_005271830.1 | 6 | 1094 | ||
XM_005271774.1 | XP_005271831.1 | 7 | 1210 | ||
XM_005271775.1 | XP_005271832.1 | 6 | 1050 | ||
XM_005271776.1 | XP_005271833.1 | 7 | 1158 | ||
XM_006715760.1 | XP_006715823.1 | 7 | 1208 | ||
XM_006715761.1 | XP_006715824.1 | 5 | 959 | ||
XM_011515461.1 | XP_011513763.1 | 7 | 1164 | ||
XM_017012466.1 | XP_016867955.1 | 8 | 1324 | ||
XM_017012467.1 | XP_016867956.1 | 8 | 1420 | ||
XM_017012468.1 | XP_016867957.1 | 6 | 1073 | ||
XM_017012469.1 | XP_016867958.1 | 6 | 1023 | ||
GenBank mRNA | AK223508.1 | - | 3 | 314 | |
AK315235.1 | - | 5 | 636 | ||
BC063614.1 | - | 6 | 872 | 3850060 | |
BX537439.1 | - | 6 | 994 |