Gene Transcripts

Gene Symbol
ACADM
Entrez Gene ID
34
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases
ACAD1, MCAD, MCADH
Chromosome Location
Chr. 1: 75724347 - 75763679 on Build GRCh38
UniGene ID
Hs.445040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000016.5 NP_000007.1 6, 7 902
NM_001127328.2 NP_001120800.1 6, 7 914
NM_001286042.1 NP_001272971.1 5, 6 814
NM_001286043.1 NP_001272972.1 7, 8 1001
GenBank mRNA AF251043.1 - 6, 7 481
AK301005.1 - 3, 4 370
AK301039.1 - 6, 7 558
AK301717.1 - 5, 6 406
AK311022.1 - 5, 6 383
AK312629.1 - 6, 7 492
AK315946.1 - 5, 6 455
BC005377.1 - 6, 7 496 3934299
BX647734.1 - 7, 8 640
KU177876.1 - 6, 7 461
KU177877.1 - 5, 6 373
KU177878.1 - 2, 3 104
M16827.1 - 6, 7 479

View Full Product Details