Gene Transcripts
- Gene Symbol
- NPC1
- Entrez Gene ID
- 4864
- Gene Name
- NPC intracellular cholesterol transporter 1
- Gene Aliases
- NPC
- Chromosome Location
- Chr. 18: 23506184 - 23586617 on Build GRCh38
- UniGene ID
- Hs.464779
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000271.4 |
NP_000262.2
|
11, 12 |
2025 |
|
XM_005258277.1 |
XP_005258334.1
|
11, 12 |
1965 |
|
XM_005258278.4 |
XP_005258335.1
|
11, 12 |
1965 |
|
XM_005258279.2 |
XP_005258336.1
|
11, 12 |
1914 |
|
XM_006722479.3 |
XP_006722542.1
|
11, 12 |
1965 |
|
XM_011526015.1 |
XP_011524317.1
|
11, 12 |
2213 |
|
XM_017025784.1 |
XP_016881273.1
|
11, 12 |
1965 |
|
XM_017025785.1 |
XP_016881274.1
|
11, 12 |
1965 |
|
XM_017025786.1 |
XP_016881275.1
|
11, 12 |
1914 |
|
XM_017025787.1 |
XP_016881276.1
|
11, 12 |
1914 |
|
GenBank mRNA |
AB209048.1 |
- |
11, 12 |
1786 |
|
AB971140.1 |
- |
11, 12 |
1751 |
|
AF002020.1 |
- |
11, 12 |
1874 |
|
AK295715.1 |
- |
9, 10 |
1665 |
|
BC063302.1 |
- |
11, 12 |
1848 |
30340517 |
View Full Product Details