Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001917.4 | NP_001908.3 | 11 | 1533 | |
XM_005268692.3 | XP_005268749.1 | 10 | 1467 | ||
XM_011538004.2 | XP_011536306.1 | 12 | 1619 | ||
XM_011538005.2 | XP_011536307.1 | 12 | 1623 | ||
GenBank mRNA | AX741655.1 | - | 11 | 1523 | |
AX741656.1 | - | 11 | 1523 | ||
AX741658.1 | - | 10 | 1408 | ||
AX741659.1 | - | 8 | 1295 | ||
AX776478.1 | - | 11 | 1523 | ||
AX776479.1 | - | 11 | 1523 | ||
AX776480.1 | - | 13 | 2452 | ||
AX786386.1 | - | 11 | 1523 | ||
BC029057.1 | - | 11 | 1503 | 5184628 | |
BM932083.1 | - | 1 | 273 | ||
BX538210.1 | - | 11 | 1465 |