Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001886.2 | NP_001877.1 | 5 | 401 | |
XM_006724140.3 | XP_006724203.1 | 5 | 459 | ||
XM_017028598.1 | XP_016884087.1 | 5 | 443 | ||
GenBank mRNA | BC069404.1 | - | 5 | 401 | |
BC096171.3 | - | 4 | 393 | 40003445 | |
BC096172.3 | - | 4 | 393 | 40003449 | |
BC096173.3 | - | 4 | 393 | 40003451 | |
BC096174.1 | - | 4 | 393 | 40003452 | |
CR456424.1 | - | 5 | 401 | ||
KM230606.1 | - | 4 | 390 | ||
KM365088.1 | - | 4 | 393 | ||
KM365089.1 | - | 5 | 401 | ||
KM365090.1 | - | 4 | 391 | ||
U59057.1 | - | 5 | 402 |