Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002336.2 | NP_002327.2 | 8 | 1887 | |
XM_006719078.3 | XP_006719141.1 | 8 | 2031 | ||
XM_011520671.2 | XP_011518973.1 | 7 | 1637 | ||
XM_017019304.1 | XP_016874793.1 | 6 | 1705 | ||
GenBank mRNA | AB209683.1 | - | 7 | 1339 | |
AF074264.1 | - | 8 | 1822 | ||
BC117136.1 | - | 8 | 1779 | 40125687 | |
BC126405.1 | - | 8 | 1779 | 8992121 | |
BC143725.1 | - | 8 | 1779 | 9052239 |