Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003420.3 | NP_003411.3 | 4 | 2486 | |
XM_017007129.1 | XP_016862618.1 | 4 | 2460 | ||
XM_017007130.1 | XP_016862619.1 | 4 | 2481 | ||
XM_017007131.1 | XP_016862620.1 | 4 | 2482 | ||
GenBank mRNA | BC013597.1 | - | 4 | 2464 | |
BC015892.1 | - | 3 | 2318 |