Gene Transcripts

Gene Symbol
NDUFA10
Entrez Gene ID
4705
Gene Name
NADH:ubiquinone oxidoreductase subunit A10
Gene Aliases
CI-42KD, CI-42k
Chromosome Location
Chr. 2: 239892450 - 240025402 on Build GRCh38
UniGene ID
Hs.277677
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322019.1 NP_001308948.1 9 1018
NM_004544.3 NP_004535.1 9 1018
NR_136155.1 8 4060
NR_136157.1 7 3891
XM_011511228.2 XP_011509530.1 9 1005
GenBank mRNA AB209440.1 - 8 4108
AF087661.1 - 9 938
AF087854.1 - 9 957
AF453834.1 - 10 1007
AK290778.1 - 9 940
BC003417.1 - 9 952 3451514

View Full Product Details