Gene Transcripts

Gene Symbol
SLC17A1
Entrez Gene ID
6568
Gene Name
solute carrier family 17 member 1
Gene Aliases
NAPI-1, NPT-1, NPT1
Chromosome Location
Chr. 6: 25723744 - 25832108 on Build GRCh38
UniGene ID
Hs.205816
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005074.3 NP_005065.2 9 1079
XM_011514818.2 XP_011513120.2 9 1116
XM_011514819.2 XP_011513121.2 8 1029
XM_011514820.2 XP_011513122.2 8 954
XM_011514821.2 XP_011513123.1 7 833
XM_017011199.1 XP_016866688.1 9 1116
XM_017011200.1 XP_016866689.1 9 1116
XM_017011201.1 XP_016866690.1 9 1116
XM_017011202.1 XP_016866691.1 8 1044
GenBank mRNA AK290783.1 - 9 1061
AK308035.1 - 9 1061
AY780791.1 - 7 801
BC101745.1 - 8 1016 8069251
BC101747.1 - 8 1016 8069253
D28532.1 - 8 960
X71355.1 - 8 975

View Full Product Details