Gene Transcripts

Gene Symbol
TCF20
Entrez Gene ID
6942
Gene Name
transcription factor 20
Gene Aliases
AR1, SPBP, TCF-20
Chromosome Location
Chr. 22: 42160013 - 42283927 on Build GRCh38
UniGene ID
Hs.475018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005650.3 NP_005641.1 2 3664
NM_181492.2 NP_852469.1 1 3563
XM_005261722.3 XP_005261779.1 2 4867
XM_006724313.3 XP_006724376.1 2 3848
XM_011530353.2 XP_011528655.1 2 3848
XM_011530354.2 XP_011528656.1 2 3848
GenBank mRNA AB006630.1 - 1 2796
KF851355.1 - 2 3664
U19345.1 - 1 283

View Full Product Details