Gene Transcripts

Gene Symbol
SCML2
Entrez Gene ID
10389
Gene Name
sex comb on midleg-like 2 (Drosophila)
Gene Aliases
Chromosome Location
Chr. X: 18239313 - 18355089 on Build GRCh38
UniGene ID
Hs.495774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006089.2 NP_006080.1 7, 8 886
NR_033717.1 7, 8 886
XM_006724458.1 XP_006724521.1 7, 8 850
XM_017029219.1 XP_016884708.1 7, 8 782
XM_017029220.1 XP_016884709.1 7, 8 778
XM_017029221.1 XP_016884710.1 7, 8 920
GenBank mRNA AK303064.1 - 6, 7 752
AK315116.1 - 7, 8 850
BC051913.1 - 7, 8 828
BC064617.1 - 7, 8 843 5575894
Y18004.1 - 7, 8 820

View Full Product Details