Gene Transcripts
- Gene Symbol
- SND1
- Entrez Gene ID
- 27044
- Gene Name
- staphylococcal nuclease and tudor domain containing 1
- Gene Aliases
- TDRD11, p100
- Chromosome Location
- Chr. 7: 127652119 - 128092607 on Build GRCh38
- UniGene ID
- Hs.122523
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014390.2 |
NP_055205.2
|
13, 14 |
1683 |
|
XM_017011987.1 |
XP_016867476.1
|
13, 14 |
1712 |
|
GenBank mRNA |
AB209510.1 |
- |
13, 14 |
1621 |
|
AK096583.1 |
- |
14, 15 |
1608 |
|
AK304181.1 |
- |
13, 14 |
1575 |
|
AK309014.1 |
- |
5, 6 |
576 |
|
AK312313.1 |
- |
13, 14 |
1637 |
|
BC017180.2 |
- |
13, 14 |
1639 |
3345037 |
BT009785.1 |
- |
13, 14 |
1457 |
|
U22055.1 |
- |
13, 14 |
1649 |
|
View Full Product Details