Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145265.1 | NP_001138737.1 | 3 | 2075 | |
NM_001145266.1 | NP_001138738.1 | 3 | 1978 | ||
NM_018389.4 | NP_060859.4 | 2 | 2349 | ||
XM_011520202.2 | XP_011518504.1 | 2 | 1236 | ||
GenBank mRNA | AK000527.1 | - | 1 | 894 | |
AK074929.1 | - | 2 | 2350 | ||
BC001427.2 | - | 2 | 1654 | 3139599 |