Gene Transcripts
- Gene Symbol
- GRHL2
- Entrez Gene ID
- 79977
- Gene Name
- grainyhead like transcription factor 2
- Gene Aliases
- BOM, DFNA28, ECTDS, TFCP2L3
- Chromosome Location
- Chr. 8: 101492440 - 101678105 on Build GRCh38
- UniGene ID
- Hs.661088
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024915.3 |
NP_079191.2
|
16 |
5068 |
|
GenBank mRNA |
AK023844.1 |
- |
16 |
4764 |
|
View Full Product Details