Gene Transcripts

Gene Symbol
CYB5B
Entrez Gene ID
80777
Gene Name
cytochrome b5 type B
Gene Aliases
CYB5-M, CYPB5M, OMB5
Chromosome Location
Chr. 16: 69424595 - 69466264 on Build GRCh38
UniGene ID
Hs.461131
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030579.2 NP_085056.2 5 1172
GenBank mRNA BC004373.1 - 5 1107
BC014431.1 - 5 1130 3542294

View Full Product Details