Gene Transcripts

Gene Symbol
TRIM4
Entrez Gene ID
89122
Gene Name
tripartite motif containing 4
Gene Aliases
RNF87
Chromosome Location
Chr. 7: 99890407 - 99919600 on Build GRCh38
UniGene ID
Hs.50749
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033017.3 NP_148977.2 7 2966
NM_033091.2 NP_149082.1 6 2888
GenBank mRNA AF220023.1 - 8 2903
AF220024.1 - 6 2825

View Full Product Details