Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193270.2 | NP_001180199.1 | 8 | 983 | |
NM_001282174.1 | NP_001269103.1 | 7 | 915 | ||
NM_006800.3 | NP_006791.2 | 7 | 968 | ||
NM_078628.1 | NP_523352.1 | 8 | 998 | ||
NM_078629.3 | NP_523353.2 | 8 | 998 | ||
XM_005274440.2 | XP_005274497.1 | 8 | 875 | ||
XM_011545442.2 | XP_011543744.1 | 8 | 952 | ||
GenBank mRNA | AF117065.1 | - | 8 | 998 | |
AK000793.1 | - | 2 | 174 | ||
AK025642.1 | - | 7 | 942 | ||
AK289780.1 | - | 8 | 920 | ||
AK294255.1 | - | 7 | 785 | ||
AK300814.1 | - | 8 | 956 | ||
AK304419.1 | - | 7 | 796 | ||
AL050178.1 | - | 1 | 47 | ||
AL713667.1 | - | 1 | 47 | ||
BC031210.1 | - | 8 | 931 | 5313214 |