Gene Transcripts

Gene Symbol
KLF14
Entrez Gene ID
136259
Gene Name
Kruppel like factor 14
Gene Aliases
BTEB5
Chromosome Location
Chr. 7: 130732554 - 130734033 on Build GRCh38
UniGene ID
Hs.125240
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138693.3 NP_619638.2 1 1238
GenBank mRNA AF490374.1 - 1 1238

View Full Product Details