Gene Transcripts

Gene Symbol
DHRSX
Entrez Gene ID
207063
Gene Name
dehydrogenase/reductase X-linked
Gene Aliases
CXorf11, DHRS5X, DHRS5Y, DHRSXY, DHRSY, SDR46C1, SDR7C6
Chromosome Location
Chr. Y: 2219506 - 2500974 on Build GRCh38
UniGene ID
Hs.131452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145177.2 NP_660160.2 7 1439
GenBank mRNA AJ293620.1 - 7 1399

View Full Product Details