Gene Transcripts
- Gene Symbol
- SLC5A12
- Entrez Gene ID
- 159963
- Gene Name
- solute carrier family 5 member 12
- Gene Aliases
- SMCT2
- Chromosome Location
- Chr. 11: 26667019 - 26723356 on Build GRCh38
- UniGene ID
- Hs.148907
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC049207.1 |
- |
9 |
1618 |
|
View Full Product Details