Gene Transcripts
- Gene Symbol
- SCML4
- Entrez Gene ID
- 256380
- Gene Name
- sex comb on midleg-like 4 (Drosophila)
- Gene Aliases
- dJ47M23.1
- Chromosome Location
- Chr. 6: 107697297 - 107845959 on Build GRCh38
- UniGene ID
- Hs.486109
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286408.1 |
NP_001273337.1
|
4, 5 |
715 |
|
NM_198081.4 |
NP_932347.2
|
5, 6 |
930 |
|
XM_011535703.2 |
XP_011534005.1
|
5, 6 |
828 |
|
XM_011535704.2 |
XP_011534006.1
|
5, 6 |
706 |
|
XM_011535706.1 |
XP_011534008.1
|
4, 5 |
672 |
|
XM_011535707.2 |
XP_011534009.1
|
5, 6 |
949 |
|
XM_017010677.1 |
XP_016866166.1
|
5, 6 |
997 |
|
XM_017010678.1 |
XP_016866167.1
|
5, 6 |
910 |
|
XM_017010679.1 |
XP_016866168.1
|
5, 6 |
808 |
|
XM_017010680.1 |
XP_016866169.1
|
5, 6 |
876 |
|
XM_017010681.1 |
XP_016866170.1
|
5, 6 |
786 |
|
XM_017010682.1 |
XP_016866171.1
|
5, 6 |
1000 |
|
XM_017010683.1 |
XP_016866172.1
|
4, 5 |
669 |
|
XM_017010684.1 |
XP_016866173.1
|
5, 6 |
1002 |
|
XM_017010685.1 |
XP_016866174.1
|
4, 5 |
796 |
|
XM_017010686.1 |
XP_016866175.1
|
4, 5 |
781 |
|
GenBank mRNA |
AK074125.1 |
- |
3, 4 |
2255 |
|
AK093571.1 |
- |
4, 5 |
715 |
|
AK303562.1 |
- |
5, 6 |
917 |
|
BC033286.1 |
- |
5, 6 |
697 |
4509724 |
CR975586.1 |
- |
4, 5 |
560 |
|
View Full Product Details