Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000848.3 | NP_000839.1 | 7 | 603 | |
NM_001142368.1 | NP_001135840.1 | 7 | 603 | ||
GenBank mRNA | AK299482.1 | - | 7 | 732 | |
AK309796.1 | - | 7 | 520 | ||
BC105038.1 | - | 7 | 534 | 8144041 | |
BC105066.1 | - | 7 | 534 | 8144069 | |
BC110380.1 | - | 7 | 556 | 5760414 | |
BM924181.1 | - | 7 | 569 | ||
BT019947.1 | - | 7 | 509 | ||
M63509.1 | - | 7 | 525 |