Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000851.3 | NP_000842.2 | 8 | 1290 | |
XM_005270785.4 | XP_005270842.1 | 5 | 1031 | ||
GenBank mRNA | AK127250.1 | - | 5 | 4098 | |
BC058881.1 | - | 8 | 1267 | 6193461 | |
BX281326.1 | - | 1 | 65 | ||
L02321.1 | - | 8 | 1274 |